6-Bromo-2-chlorobenzothiazole ≥96.0%

Leverantör: TCI
Varning

B4448-1G B4448-5G
TCIAB4448-1GEA 605 SEK
TCIAB4448-1G TCIAB4448-5G
6-Bromo-2-chlorobenzothiazole ≥96.0%
6-Bromo-2-chlorobenzothiazole
Formel: C₇H₃BrClNS
Kokpunkt: 157°C
MDL Number: MFCD04971822
CAS nummer: 80945-86-4

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR