2-Bromotriphenylene ≥98.0%

Leverantör: TCI
Varning

B4442-1G B4442-5G
TCIAB4442-1GEA 710 SEK
TCIAB4442-1G TCIAB4442-5G
2-Bromotriphenylene ≥98.0%
2-Bromotriphenylene
Formel: C₁₈H₁₁Br
MDL Number: MFCD16659043
CAS nummer: 19111-87-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR