2-Bromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
Leverantör: TCI
|
Varning
|
Synonyms:
2-Bromo-9-phenyl-9H-carbazole
B4439-1G
B4439-5G
TCIAB4439-1GEA
605
SEK
TCIAB4439-1G
TCIAB4439-5G
2-Bromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
2-Bromo-9-phenylcarbazole
Formel:
C₁₈H₁₂BrN MW: 322.21 g/mol |
MDL Number:
MFCD24387061 CAS nummer: 94994-62-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...