2,2-Bis(3-amino-4-hydroxylphenyl)propane ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4,4'-Isopropylidenebis(2-aminophenol), 4,4'-(Propane-2,2-diyl)bis(2-aminophenol)

B4429-5G B4429-25G
TCIAB4429-5GEA 405 SEK
TCIAB4429-5G TCIAB4429-25G
2,2-Bis(3-amino-4-hydroxylphenyl)propane ≥97.0% (by HPLC, titration analysis)
2,2-Bis(3-amino-4-hydroxylphenyl)propane
Formel: C₁₅H₁₈N₂O₂
MW: 258,32 g/mol
MDL Number: MFCD00437413
CAS nummer: 1220-78-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR