(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(1R,2R)-(-)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene), (R,R)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine, (R,R)-Jacobsen Ligand
B4418-25G
B4418-5G
TCIAB4418-25GEA
3575
SEK
TCIAB4418-25G
TCIAB4418-5G
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine ≥98.0% (by HPLC, total nitrogen)
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine
Formel:
C₃₆H₅₄N₂O₂ MW: 546,84 g/mol |
MDL Number:
MFCD00191800 CAS nummer: 135616-40-9 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 206.0 to 210.0 deg-C |
Specific rotation [a]20/D | -310 to -330 deg (C=1,CH2Cl2) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...