6-Bromo-2H-1,4-benzoxazin-3(4H)-one ≥94.0%
Leverantör: TCI EUROPE
|
Varning
|
B4402-1G
B4402-5G
TCIAB4402-1GEA
550
SEK
TCIAB4402-1G
TCIAB4402-5G
6-Bromo-2H-1,4-benzoxazin-3(4H)-one ≥94.0%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one
Formel:
C₈H₆BrNO₂ MW: 228,04 g/mol |
MDL Number:
MFCD00461173 CAS nummer: 24036-52-0 |
Specifikation testresultat
Purity (GC) | min. 94.0 % |
Melting point | 219.0 to 223.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...