2,3-Bis(4-hydroxyphenyl)propionitrile ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: DPN
B4385-10MG B4385-50MG
TCIAB4385-10MGEA 685 SEK
TCIAB4385-10MG TCIAB4385-50MG
2,3-Bis(4-hydroxyphenyl)propionitrile ≥98.0%
2,3-Bis(4-hydroxyphenyl)propionitrile
Formel: C₁₅H₁₃NO₂
MW: 239,27 g/mol
Storage Temperature: Frys
MDL Number: MFCD01695412
CAS nummer: 1428-67-7
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 200.0 to 204.0 deg-C