Bis(2-hydroxyethyl)dimethylammonium chloride 98,0 Titrimetrisk

Leverantör: TCI
Varning

B4384-5G B4384-25G
TCIAB4384-5GEA 485 SEK
TCIAB4384-5G TCIAB4384-25G
Bis(2-hydroxyethyl)dimethylammonium chloride 98,0 Titrimetrisk
Bis(2-hydroxyethyl)dimethylammonium chloride
Formel: C₆H₁₆ClNO₂
MW: 169.65 g/mol
MDL Number: MFCD00041987
CAS nummer: 38402-02-7
EINECS: 253-913-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR