3-Bromopyridine-2-carbonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Bromopicolinonitrile, 3-Bromo-2-cyanopyridine
B4382-1G
B4382-5G
TCIAB4382-1GEA
565
SEK
TCIAB4382-1G
TCIAB4382-5G
3-Bromopyridine-2-carbonitrile ≥98.0%
3-Bromopyridine-2-carbonitrile
Formel:
C₆H₃BrN₂ MW: 183,01 g/mol Kokpunkt: 120°C |
MDL Number:
MFCD02683288 CAS nummer: 55758-02-6 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...