4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-S-indacene ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
EMind-Br
B4379-1G
B4379-5G
TCIAB4379-1GEA
1050
SEK
TCIAB4379-1G
TCIAB4379-5G
4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-S-indacene ≥95.0%
4-Bromo-1,1,7,7-tetraethyl-1,2,3,5,6,7-hexahydro-3,3,5,5-tetramethyl-S-indacene
Formel:
C₂₄H₃₇Br MW: 405,46 g/mol |
CAS nummer:
1142818-90-3 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...