Bismuth tripotassium dicitrate

Leverantör: TCI

Synonyms: Bismuth(III) tripotassium 2-hydroxypropane-1,2,3-tricarboxylate, Bismuth potassium citrate

B4377-1G B4377-5G
TCIAB4377-1GEA 485 SEK
TCIAB4377-1G TCIAB4377-5G
Bismuth tripotassium dicitrate
Bismuth tripotassium dicitrate
Formel: C₁₂H₁₀BiK₃O₁₄
MW: 704.49 g/mol
MDL Number: MFCD02101411
CAS nummer: 57644-54-9
EINECS: 200-872-5
Merck Index: 14,02485

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystaline
Bismuth 30.0 to 36.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR