1-(2-Bromoethoxy)-4-fluorobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Fluorophenoxy-ethylbromide

B4376-5G B4376-25G
TCIAB4376-5GEA 1275 SEK
TCIAB4376-5G TCIAB4376-25G
1-(2-Bromoethoxy)-4-fluorobenzene ≥98.0%
1-(2-Bromoethoxy)-4-fluorobenzene
Formel: C₈H₈BrFO
Kokpunkt: 66°C
MDL Number: MFCD00044739
CAS nummer: 332-48-9

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR