1-Bromo-4-chloro-3-nitrobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Bromo-1-chloro-2-nitrobenzene, 5-Bromo-2-chloronitrobenzene

B4374-5G B4374-25G
TCIAB4374-5GEA 325 SEK
TCIAB4374-5G TCIAB4374-25G
1-Bromo-4-chloro-3-nitrobenzene ≥98.0%
1-Bromo-4-chloro-3-nitrobenzene
Formel: C₆H₃BrClNO₂
MW: 236,4500 g/mol
MDL Number: MFCD05863216
CAS nummer: 16588-24-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR