1,3-Benzenedithiol ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Benzene-1,3-dithiol
B4371-1G B4371-5G
TCIAB4371-1GEA 1225 SEK
TCIAB4371-1G TCIAB4371-5G
1,3-Benzenedithiol ≥95.0%
1,3-Benzenedithiol
Formel: C₆H₆S₂
MW: 142,25 g/mol
Kokpunkt: 245°C
Smältpunkt: 27°C
MDL Number: MFCD00004842
CAS nummer: 626-04-0
EINECS: 210-925-3
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %