5-Bromo-2-fluoro-m-xylene ≥97.0%
Leverantör: TCI EUROPE
Varning
B4370-1G B4370-5G
TCIAB4370-1GEA 390 SEK
TCIAB4370-1G TCIAB4370-5G
5-Bromo-2-fluoro-m-xylene ≥97.0%
5-Bromo-2-fluoro-m-xylene
Formel: C₈H₈BrF
MW: 203,05 g/mol
Kokpunkt: 89°C
MDL Number: MFCD01320701
CAS nummer: 99725-44-7
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %