1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(Phenylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
B4364-1G
B4364-5G
TCIAB4364-1GEA
775
SEK
TCIAB4364-1G
TCIAB4364-5G
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0%
1-Benzyl-1H-pyrazole-4-boronic acid pinacol ester
Formel:
C₁₆H₂₁BN₂O₂ MW: 284,16 g/mol |
MDL Number:
MFCD03789252 CAS nummer: 761446-45-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...