3-Bromo-5-iodopyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

B4346-1G B4346-5G
TCIAB4346-1GEA 1150 SEK
TCIAB4346-1G TCIAB4346-5G
3-Bromo-5-iodopyridine ≥98.0%
3-Bromo-5-iodopyridine
Formel: C₅H₃BrIN
MW: 283,89 g/mol
MDL Number: MFCD03086019
CAS nummer: 233770-01-9

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR