N-tert-Butyl-2-thiophenesulphonamide ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-(tert-Butylaminosulphonyl)thiophene
B4330-1G
B4330-5G
TCIAB4330-1GEA
350
SEK
TCIAB4330-1G
TCIAB4330-5G
N-tert-Butyl-2-thiophenesulphonamide ≥98.0%
N-tert-Butyl-2-thiophenesulphonamide
Formel:
C₈H₁₃NO₂S₂ MW: 219,32 g/mol |
MDL Number:
MFCD02047252 CAS nummer: 100342-30-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...