(Bromodifluoromethyl)trimethylsilane ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Bromodifluoro(trimethylsilyl)methane

B4325-5G B4325-1G
TCIAB4325-5GEA 2775 SEK
TCIAB4325-5G TCIAB4325-1G
(Bromodifluoromethyl)trimethylsilane ≥98.0%
(Bromodifluoromethyl)trimethylsilane
Formel: C₄H₉BrF₂Si
MW: 203,10 g/mol
Kokpunkt: 108°C
MDL Number: MFCD18641931
CAS nummer: 115262-01-6
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa4158f1f214035bd0ec0d1711d348a6fe86ea07&vid=aa4158f1f214035bd0ec0d1711d348a6fe86ea07&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR