1-(2-Bromoethoxy)-4-chlorobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Chloro-ω-bromophenetol, 4-Chlorophenyl 2-bromoethyl ether, 4-Chloro-β-bromophenetol
B4298-5G
B4298-25G
TCIAB4298-5GEA
1950
SEK
TCIAB4298-5G
TCIAB4298-25G
1-(2-Bromoethoxy)-4-chlorobenzene ≥98.0%
1-(2-Bromoethoxy)-4-chlorobenzene
Formel:
C₈H₈BrClO MW: 235,51 g/mol Kokpunkt: 87°C |
MDL Number:
MFCD00000617 CAS nummer: 2033-76-3 EINECS: 217-994-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...