2,2'-Bipyrazine ≥97.0%
Leverantör: TCI
|
Varning
|
B4297-100MG
B4297-500MG
TCIAB4297-100MGEA
1100
SEK
TCIAB4297-100MG
TCIAB4297-500MG
2,2'-Bipyrazine ≥97.0%
2,2'-Bipyrazine
Formel:
C₈H₆N₄ |
MDL Number:
MFCD00010288 CAS nummer: 10199-00-5 |
|
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 185.0 to 189.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...