N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Perylene Orange

B4268-1G B4268-5G
TCIAB4268-1GEA 4600 SEK
TCIAB4268-1G TCIAB4268-5G
N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥98.0%
N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide
Formel: C₄₈H₄₂N₂O₄
MW: 710,87 g/mol
CAS nummer: 82953-57-9

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR