tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Norbornene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]-5-heptene-2-carboxylate

B4267-5G B4267-25G
TCIAB4267-5GEA 495 SEK
TCIAB4267-5G TCIAB4267-25G
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) ≥95.0%
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture)
Formel: C₁₂H₁₈O₂
MW: 194,27 g/mol
Kokpunkt: 103°C
MDL Number: MFCD18083704
CAS nummer: 154970-45-3

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR