Bendazac ≥97.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
[(1-Benzyl-3-indazolyl)oxy]acetic Acid, Bendazolic acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole
B4223-1G
B4223-5G
TCIAB4223-1GEA
550
SEK
TCIAB4223-1G
TCIAB4223-5G
Bendazac ≥97.0% (by GC, titration analysis)
Bendazac
Formel:
C₁₆H₁₄N₂O₃ |
MDL Number:
MFCD00866158 CAS nummer: 20187-55-7 EINECS: 243-569-2 Merck Index: 14,01035 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...