4-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
|
Varning
|
B4216-5G
B4216-25G
TCIAB4216-5GEA
335
SEK
TCIAB4216-5G
TCIAB4216-25G
4-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
4-Bromobenzamide
Formel:
C₇H₆BrNO |
MDL Number:
MFCD00007991 CAS nummer: 698-67-9 EINECS: 211-817-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 191.0 to 195.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...