1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,4-Di([2,2':6',2''-terpyridin]-4'-yl)benzene
B4194-200MG
B4194-1G
TCIAB4194-200MGEA
885
SEK
TCIAB4194-200MG
TCIAB4194-1G
1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene ≥97.0%
1,4-Di[[2,2':6',2''-terpyridin]-4'-yl]benzene
Formel:
C₃₆H₂₄N₆ MW: 540,63 g/mol Smältpunkt: 250°C |
MDL Number:
MFCD02093699 CAS nummer: 146406-75-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...