4-Bromobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Bromophenylamidine hydrochloride, 4-Bromobenzimidamide hydrochloride, 4-Bromobenzene-1-carboximidamide hydrochloride
B4161-1G
B4161-5G
TCIAB4161-1GEA
375
SEK
TCIAB4161-1G
TCIAB4161-5G
4-Bromobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Bromobenzamidine hydrochloride
Formel:
C₇H₈BrClN₂ MW: 235,51 g/mol |
MDL Number:
MFCD00464967 CAS nummer: 55368-42-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...