7-Bromoquinoline ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

B4155-1G B4155-5G
TCIAB4155-1GEA 1050 SEK
TCIAB4155-1G TCIAB4155-5G
7-Bromoquinoline ≥98.0% (by GC, titration analysis)
7-Bromoquinoline
Formel: C₉H₆BrN
MW: 208,06 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD03695823
CAS nummer: 4965-36-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 34.0 to 38.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR