4-Bromo-2-nitrobenzaldehyde 96,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
B4146-1G
B4146-5G
TCIAB4146-1GEA
590
SEK
TCIAB4146-1G
TCIAB4146-5G
4-Bromo-2-nitrobenzaldehyde 96,0 GC_ANALYSMETOD
4-Bromo-2-nitrobenzaldehyde
Formel:
C₇H₄BrNO₃ MW: 230.0156 g/mol |
MDL Number:
MFCD00463687 CAS nummer: 5551-12-2 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...