4-Bromothiophene-3-carbaldehyde ≥97.0%
Leverantör: TCI EUROPE
B4126-200MG
B4126-1G
TCIAB4126-200MGEA
1075
SEK
TCIAB4126-200MG
TCIAB4126-1G
4-Bromothiophene-3-carbaldehyde ≥97.0%
4-Bromothiophene-3-carbaldehyde
Formel:
C₅H₃BrOS MW: 191,05 g/mol Kokpunkt: 120°C |
MDL Number:
MFCD01318960 CAS nummer: 18791-78-1 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...