(S)-(+)-1-Benzyloxy-2-propanol ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (S)-1-Benzyloxy-2-propanol, Benzyl (S)-(+)-2-Hydroxypropyl Ether
B4117-1G B4117-5G
TCIAB4117-1GEA 710 SEK
TCIAB4117-1G TCIAB4117-5G
(S)-(+)-1-Benzyloxy-2-propanol ≥95.0%
(S)-(+)-1-Benzyloxy-2-propanol
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
Kokpunkt: 81°C
Flampunkt: 108°C
MDL Number: MFCD07367008
CAS nummer: 85483-97-2
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D +12.0 to +15.0 deg (C=1、CHCL3)