6-Bromo-2,2'-bipyridyl ≥97.0%

Leverantör: TCI
Varning

Synonyms: 6-Bromo-2,2'-bipyridine

B4114-1G B4114-5G
TCIAB4114-1GEA 1725 SEK
TCIAB4114-1G TCIAB4114-5G
6-Bromo-2,2'-bipyridyl ≥97.0%
6-Bromo-2,2'-bipyridyl
Formel: C₁₀H₇BrN₂
Kokpunkt: 133°C
MDL Number: MFCD01318953
CAS nummer: 10495-73-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR