(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (S)-4-(tert-Butyl)-2-(2-pyridyl)-4,5-dihydrooxazole
B4104-1G B4104-5G
TCIAB4104-1GEA 1550 SEK
TCIAB4104-1G TCIAB4104-5G
(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline ≥98.0%
(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline
Formel: C₁₂H₁₆N₂O
MW: 204,27 g/mol
MDL Number: MFCD22200533
CAS nummer: 117408-98-7
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C
Specific rotation [a]20/D -75.0 to -79.0 deg (C=1, CH2CL2)