6-Bromo-1H-indazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B4080-1G
B4080-5G
B4080-25G
TCIAB4080-1GEA
255
SEK
TCIAB4080-1G
TCIAB4080-5G
TCIAB4080-25G
6-Bromo-1H-indazole ≥98.0%
6-Bromo-1H-indazole
Formel:
C₇H₅BrN₂ MW: 197,0300 g/mol |
MDL Number:
MFCD03265457 CAS nummer: 79762-54-2 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 181.0 to 185.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...