Methyl palmitate 95%
Leverantör: Thermo Scientific
Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
414681000 414685000
ACRO414681000EA 465 SEK
ACRO414681000 ACRO414685000
Methyl palmitate 95%
Methyl palmitate
Formel: C₁₇H₃₄O₂
MW: 270,46 g/mol
Kokpunkt: 417 °C (1013 hPa)
Smältpunkt: 28 °C
Densitet: 0,8804 g/cm³ (20 °C)
Flampunkt: >110 °C
MDL Number: MFCD00008994
CAS nummer: 112-39-0
EINECS: 203-966-3
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %