1-Butylpiperazine ≥98.0%

Leverantör: TCI
Varning

B4078-5ML B4078-25ML
TCIAB4078-5MLEA 325 SEK
TCIAB4078-5ML TCIAB4078-25ML
1-Butylpiperazine ≥98.0%
1-Butylpiperazine
Formel: C₈H₁₈N₂
Kokpunkt: 192°C
MDL Number: MFCD00039702
CAS nummer: 5610-49-1

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR