3-Bromo-2-chloro-5-nitropyridine ≥98.0%

Leverantör: TCI
Varning

B4077-1G B4077-5G
TCIAB4077-1GEA 415 SEK
TCIAB4077-1G TCIAB4077-5G
3-Bromo-2-chloro-5-nitropyridine ≥98.0%
3-Bromo-2-chloro-5-nitropyridine
Formel: C₅H₂BrClN₂O₂
MDL Number: MFCD00233989
CAS nummer: 5470-17-7

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR