5-Bromo-2-methox-4-methylypyridine ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 5-Bromo-2-methoxy-4-methylpyridine
B4067-1G B4067-5G
TCIAB4067-1GEA 375 SEK
TCIAB4067-1G TCIAB4067-5G
5-Bromo-2-methox-4-methylypyridine ≥98.0%
5-Bromo-2-methox-4-methylypyridine
Formel: C₇H₈BrNO
MW: 202,0485 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD04039980
CAS nummer: 164513-39-7
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C