2-Butylimidazole-4-carbaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Butyl-1H-imidazole-5-carboxaldehyde

B4064-5G B4064-25G
TCIAB4064-5GEA 1125 SEK
TCIAB4064-5G TCIAB4064-25G
2-Butylimidazole-4-carbaldehyde ≥98.0%
2-Butylimidazole-4-carbaldehyde
Formel: C₈H₁₂N₂O
MW: 152,193 g/mol
MDL Number: MFCD01075030
CAS nummer: 68282-49-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR