N-Boc-2-pyrroleboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1H-Pyrrole-2-boronic acid, N-BOC protected, N-Boc-2-Pyrroleboronic acid
B4063-1G
B4063-5G
TCIAB4063-1GEA
925
SEK
TCIAB4063-1G
TCIAB4063-5G
N-Boc-2-pyrroleboronic acid (contains varying amounts of Anhydride)
N-Boc-2-pyrroleboronic acid
Formel:
C₉H₁₄BNO₄ MW: 211,02 g/mol Smältpunkt: 102°C Storage Temperature: Kylskåp |
MDL Number:
MFCD01318939 CAS nummer: 135884-31-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...