N-Benzyltropinone ≥95.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one, (1R,5S)-8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

B4058-1G B4058-5G
TCIAB4058-1GEA 245 SEK
TCIAB4058-1G TCIAB4058-5G
N-Benzyltropinone ≥95.0% (by GC, titration analysis)
N-Benzyltropinone
Formel: C₁₄H₁₇NO
MW: 215,30 g/mol
Kokpunkt: 124°C
MDL Number: MFCD00151640
CAS nummer: 28957-72-4
EINECS: 249-329-3

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR