3'-Bromo-2,2':5',2''-terthiophene ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 3-Bromo-2,5-di(2-thienyl)thiophene
B4050-1G B4050-5G
TCIAB4050-1GEA 4625 SEK
TCIAB4050-1G TCIAB4050-5G
3'-Bromo-2,2':5',2''-terthiophene ≥97.0%
3'-Bromo-2,2':5',2''-terthiophene
Formel: C₁₂H₇BrS₃
MW: 327,27 g/mol
CAS nummer: 105125-00-6
Order Now
Specifikation testresultat
Appearance White or Colorless to Amber to Dark green powder to lump to clear liquid
Purity (GC) min. 97.0 %