Boc-L-homoserine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: (S)-2-(Boc-amino)-4-hydroxybutyric acid, Boc-L-Homoserine, (S)-2-(tert-Butoxycarbonylamino)-4-hydroxybutyric acid, N-Boc-L-homoserine, N-(tert-Butoxycarbonyl)-L-homoserine

B4025-1G B4025-5G
TCIAB4025-1GEA 485 SEK
TCIAB4025-1G TCIAB4025-5G
Boc-L-homoserine ≥98.0% (by HPLC, titration analysis)
Boc-L-homoserine
Formel: C₉H₁₇NO₅
MW: 219,2350 g/mol
Smältpunkt: 140°C
Storage Temperature: Frys
MDL Number: MFCD00057839
CAS nummer: 41088-86-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -28.0 to -32.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR