3-(tert-Butyl)-2-hydroxybenzaldehyde ≥95.0%
Leverantör: TCI
|
Varning
|
B4016-1G
B4016-5G
TCIAB4016-1GEA
310
SEK
TCIAB4016-1G
TCIAB4016-5G
3-(tert-Butyl)-2-hydroxybenzaldehyde ≥95.0%
3-(tert-Butyl)-2-hydroxybenzaldehyde
Formel:
C11H14O2 Kokpunkt: 79°C |
MDL Number:
MFCD00778877 CAS nummer: 24623-65-2 |
Specifikation testresultat
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...