tert-Butyl-3-oxoazetidine-1-carboxylate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N-Boc-3-azetidinone, Tert-butyl 3-oxoazetidine-1-carboxylate, tert-Butyl 3-oxoazetidine-1-carboxylate, 1-BOC-3-Azetidinone

B3988-1G B3988-5G
TCIAB3988-1GEA 790 SEK
TCIAB3988-1G TCIAB3988-5G
tert-Butyl-3-oxoazetidine-1-carboxylate ≥98.0%
tert-Butyl-3-oxoazetidine-1-carboxylate
Formel: C₈H₁₃NO₃
MW: 171,2 g/mol
MDL Number: MFCD01861741
CAS nummer: 398489-26-4

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR