1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane ≥96.0%

Leverantör: TCI
Varning

B3973-200MG B3973-1G
TCIAB3973-200MGEA 775 SEK
TCIAB3973-200MG TCIAB3973-1G
1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane ≥96.0%
1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane
Formel: C₂₄H₂₂N₄
CAS nummer: 96897-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR