1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct adduct ≥94.0%

Leverantör: TCI EUROPE
Varning

Synonyms: DABSO, 1,4-Diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulphinate, DABCO-Bis(sulphur dioxide), Bis(sulphur dioxide)-1,4-diazabicyclo[2.2.2]octane

B3960-1G B3960-5G
TCIAB3960-1GEA 350 SEK
TCIAB3960-1G TCIAB3960-5G
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct adduct ≥94.0%
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct
Formel: C₆H₁₂N₂O₄S₂
MW: 240,29 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22377802
CAS nummer: 119752-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad407288f27f9bb7715c81410c9a664112708a7&vid=7ad407288f27f9bb7715c81410c9a664112708a7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Iodometric back titration) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR