2-Bromo-7-tert-butylpyrene ≥98.0%

Leverantör: TCI EUROPE

B3957-1G B3957-5G
TCIAB3957-1GEA 4100 SEK
TCIAB3957-1G TCIAB3957-5G
2-Bromo-7-tert-butylpyrene ≥98.0%
2-Bromo-7-tert-butylpyrene
Formel: C₂₀H₁₇Br
MW: 337,26 g/mol
CAS nummer: 78751-80-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR