Ethyl palmitate ≥97%
Leverantör: Thermo Scientific
Synonyms: Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester
A15694.14 A15694.22 A15694.36
A15694.14EA 1300 SEK
A15694.14 A15694.22 A15694.36
Ethyl palmitate ≥97%
Ethyl palmitate
Purity: 97% CAS No.: 628-97-7 Molecular Formula: CH3(CH2)14CO2CH2CH3 Formula Weight: 284.48 Flash Point: 178°(352°F) Refractive Index: 1.4395 Melting Point: 22-26° Boiling Point: 192-193°/10mm Beilstein Reference: 1782663 MDL No.: MFCD00008996
Formel: C₁₈H₃₆O₂
MW: 284,48 g/mol
Kokpunkt: 192…193 °C (10 mmHg)
Smältpunkt: 22…26 °C
Densitet: 0,857
Flampunkt: 178 °C (352 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008996
CAS nummer: 628-97-7
EINECS: 211-064-6