5-Bromo-2-chlorobenzyl alcohol ≥98.0%
Leverantör: TCI
Synonyms:
(5-Bromo-2-chlorophenyl)methanol
B3946-1G
B3946-5G
TCIAB3946-1GEA
575
SEK
TCIAB3946-1G
TCIAB3946-5G
5-Bromo-2-chlorobenzyl alcohol ≥98.0%
5-Bromo-2-chlorobenzyl alcohol
Formel:
C₇H₆BrClO MW: 221.48 g/mol |
MDL Number:
MFCD02683548 CAS nummer: 149965-40-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...